Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppcs
Entrez Gene ID
561272
Gene Name
phosphopantothenoylcysteine synthetase
Gene Aliases
im:7147515, zgc:136566
Chromosome Location
Chr.11: 26363438 - 26369549 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045240.1 NP_001038705.1 3 - 4 769 96
GenBank mRNA BC116589.1 - 3 - 4 769 96
BC153531.1 - 3 - 4 726 96

View Full Product Details