Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc2a11l
Entrez Gene ID
564849
Gene Name
solute carrier family 2 member 11, like
Gene Aliases
zgc:158393
Chromosome Location
Chr.10: 17485945 - 17501505 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080016.1 NP_001073485.1 8 - 9 996 92
GenBank mRNA BC128857.1 - 8 - 9 1019 92

View Full Product Details