Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taar14l
Entrez Gene ID
100034645
Gene Name
trace amine associated receptor 14l
Gene Aliases
si:dkey-246e3.2
Chromosome Location
Chr.20: 46245450 - 46246407 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083103.1 NP_001076572.1 1 - 1 445 117

View Full Product Details