Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lmo4a
Entrez Gene ID
114412
Gene Name
LIM domain only 4a
Gene Aliases
NH13, id:ibd2576, id:ibd5151, lmo4
Chromosome Location
Chr.5: 24454 - 46040 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177984.1 NP_817093.1 2 - 3 611 75
XM_009301446.3 XP_009299721.1 2 - 3 629 75
XM_017356026.2 XP_017211515.1 3 - 4 1508 75
GenBank mRNA AY028903.1 - 2 - 3 599 75
BC045835.1 - 2 - 3 611 75
BC164026.1 - 1 - 2 270 75

View Full Product Details