Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adkb
Entrez Gene ID
322229
Gene Name
adenosine kinase b
Gene Aliases
cb980, fb53h03, wu:fb53h03
Chromosome Location
Chr.12: 46842395 - 46868003 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198802.1 NP_942097.1 8 - 9 804 56
GenBank mRNA BC051621.1 - 8 - 9 804 56

View Full Product Details