Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
naa15a
Entrez Gene ID
323672
Gene Name
N-alpha-acetyltransferase 15, NatA auxiliary subunit a
Gene Aliases
narg1a, wu:fc06f11, wu:fc23c05, zgc:66136
Chromosome Location
Chr.14: 46118834 - 46155077 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200646.2 NP_956940.2 9 - 10 1219 135
GenBank mRNA BC045491.1 - 9 - 10 1188 135
BC057466.1 - 9 - 10 1219 135
BC165243.1 - 9 - 10 1052 135

View Full Product Details