Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prkg1a
Entrez Gene ID
394005
Gene Name
protein kinase cGMP-dependent 1a
Gene Aliases
prkg1, zgc:63955
Chromosome Location
Chr.13: 40815015 - 40955692 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201030.1 NP_957324.1 8 - 9 1235 85
XM_005157058.4 XP_005157115.1 7 - 8 1269 85
GenBank mRNA BC054581.1 - 8 - 9 1235 85

View Full Product Details