Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
actr5
Entrez Gene ID
406809
Gene Name
actin related protein 5
Gene Aliases
wu:fi39h10, zgc:63743
Chromosome Location
Chr.9: 331487 - 340294 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213488.1 NP_998653.1 6 - 7 1250 68
GenBank mRNA BC055228.1 - 6 - 7 1250 68

View Full Product Details