Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nus1
Entrez Gene ID
436629
Gene Name
NUS1 dehydrodolichyl diphosphate synthase subunit
Gene Aliases
fc21g03, wu:fc21g03, zgc:92136
Chromosome Location
Chr.20: 42223824 - 42241429 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002356.1 NP_001002356.1 2 - 3 617 99
GenBank mRNA BC075899.1 - 2 - 3 617 99

View Full Product Details