Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aicda
Entrez Gene ID
493784
Gene Name
activation-induced cytidine deaminase
Gene Aliases
-
Chromosome Location
Chr.16: 12660477 - 12665652 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008403.1 NP_001008403.1 3 - 4 512 98
GenBank mRNA AY528720.1 - 3 - 4 512 98
BC162573.1 - 3 - 4 488 98

View Full Product Details