Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
drd4a
Entrez Gene ID
503564
Gene Name
dopamine receptor D4a
Gene Aliases
zgc:158147
Chromosome Location
Chr.25: 4707173 - 4733221 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012616.3 NP_001012634.2 1 - 2 463 80
GenBank mRNA AY750152.1 - 1 - 2 250 80
BC129139.1 - 1 - 2 463 80

View Full Product Details