Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atpaf1
Entrez Gene ID
550606
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Gene Aliases
id:ibd1136, si:dkey-171o17.2, zgc:110583
Chromosome Location
Chr.22: 17192769 - 17201398 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017907.2 NP_001017907.2 1 - 2 207 92
GenBank mRNA BC092940.1 - 1 - 2 208 92

View Full Product Details