Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rpain
Entrez Gene ID
751671
Gene Name
RPA interacting protein
Gene Aliases
zgc:153251
Chromosome Location
Chr.5: 3891470 - 3908849 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045388.1 NP_001038853.1 3 - 4 404 114
XM_005171732.4 XP_005171789.1 3 - 4 457 114
GenBank mRNA BC122206.1 - 3 - 4 404 114

View Full Product Details