Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ckba
Entrez Gene ID
564694
Gene Name
creatine kinase, brain a
Gene Aliases
cb02h12, fb62d04, fc14g11, mg:cb02h12, wu:fb62d04, wu:fc14g11, zgc:154095, zgc:92079
Chromosome Location
Chr.13: 15682085 - 15700112 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077163.1 NP_001070631.1 4 - 5 614 97
XM_005156643.4 XP_005156700.1 4 - 5 634 97
XM_017358772.2 XP_017214261.1 3 - 4 797 97
GenBank mRNA BC085630.1 - 4 - 5 585 97
BC124538.1 - 4 - 5 614 97

View Full Product Details