Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ccna2
Entrez Gene ID
192295
Gene Name
cyclin A2
Gene Aliases
CycA2, cb671, chunp6924, wu:fi36f03
Chromosome Location
Chr.14: 46357824 - 46374670 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152949.1 NP_694481.1 2 - 3 453 125
XM_021480961.1 XP_021336636.1 1 - 2 561 125
GenBank mRNA AF234784.2 - 2 - 3 453 125
BC045840.1 - 2 - 3 453 125
BC068323.1 - 2 - 3 268 125
BC151890.1 - 2 - 3 448 125

View Full Product Details