Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdcd10b
Entrez Gene ID
405869
Gene Name
programmed cell death 10b
Gene Aliases
zgc:85629
Chromosome Location
Chr.2: 36721459 - 36731963 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212933.1 NP_998098.1 7 - 8 678 115
GenBank mRNA BC067550.1 - 7 - 8 678 115

View Full Product Details