Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a28
Entrez Gene ID
100535877
Gene Name
solute carrier family 25 member 28
Gene Aliases
mfrn2, wu:fc48b02, wu:fc66h02, zgc:64212
Chromosome Location
Chr.13: 27345 - 46895 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213119.2 NP_998284.2 3 - 4 807 68
GenBank mRNA BC054641.1 - 3 - 4 749 68
EH536337.1 - 3 - 4 801 68

View Full Product Details