Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gspt1
Entrez Gene ID
445485
Gene Name
G1 to S phase transition 1
Gene Aliases
zgc:91975
Chromosome Location
Chr.12: 598232 - 619798 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003992.1 NP_001003992.1 9 - 10 1299 69
GenBank mRNA BC080263.1 - 9 - 10 1299 69
BC164587.1 - 9 - 10 1165 69

View Full Product Details