The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006017.1 | NP_001006017.1 | 1 - 2 | 374 | 90 |
GenBank mRNA | BC083406.1 | - | 1 - 2 | 374 | 90 |
BC163994.1 | - | 1 - 2 | 253 | 90 |