Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cpox
Entrez Gene ID
321294
Gene Name
coproporphyrinogen oxidase
Gene Aliases
cb535, cpo, im:7156501, sb:cb535, wu:fc43a01
Chromosome Location
Chr.8: 18601704 - 18613987 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040094.2 NP_001035183.2 1 - 2 587 72
GenBank mRNA AB180841.1 - 1 - 2 599 72
BC124459.1 - 1 - 2 587 72

View Full Product Details