Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ctdsplb
Entrez Gene ID
767676
Gene Name
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like b
Gene Aliases
wu:fd18d04, zgc:154102
Chromosome Location
Chr.2: 21356199 - 21383393 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076615.1 NP_001070083.1 4 - 5 715 72
XM_005163361.4 XP_005163418.1 5 - 6 1022 72
GenBank mRNA BC124542.1 - 4 - 5 715 72

View Full Product Details