Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem51a
Entrez Gene ID
777622
Gene Name
transmembrane protein 51a
Gene Aliases
im:7148815, wu:fj97e11, zgc:152859
Chromosome Location
Chr.8: 317393 - 326664 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077730.1 NP_001071198.1 2 - 3 634 93
GenBank mRNA BC125824.1 - 2 - 3 634 93

View Full Product Details