Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sephs1
Entrez Gene ID
324947
Gene Name
selenophosphate synthetase 1
Gene Aliases
wu:fc49b09, zgc:55304
Chromosome Location
Chr.18: 130 - 8135 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199701.2 NP_955995.2 7 - 8 1005 81
XM_005159006.4 XP_005159063.1 7 - 8 906 81
GenBank mRNA BC045302.1 - 7 - 8 863 81
BC164487.1 - 6 - 7 783 81

View Full Product Details