Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gipc2
Entrez Gene ID
393904
Gene Name
GIPC PDZ domain containing family, member 2
Gene Aliases
gipc1, rgs19ip1, zgc:56157, zgc:77689
Chromosome Location
Chr.2: 9560711 - 9590849 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200930.1 NP_957224.1 4 - 5 974 68
GenBank mRNA AY989814.1 - 4 - 5 754 68
BC045953.1 - 4 - 5 974 68
BC064297.1 - 4 - 5 68
BC155746.1 - 4 - 5 68

View Full Product Details