Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp3c1
Entrez Gene ID
324340
Gene Name
cytochrome P450, family 3, subfamily c, polypeptide 1
Gene Aliases
CYP3A, wu:fa96d12, wu:fc26b04, zgc:56626
Chromosome Location
Chr.3: 40969242 - 40976532 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212673.1 NP_997838.1 5 - 6 431 118
GenBank mRNA AY499002.1 - 4 - 5 371 118
BC052130.1 - 5 - 6 431 118

View Full Product Details