Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cul1b
Entrez Gene ID
406816
Gene Name
cullin 1b
Gene Aliases
cul1, wu:fi43h07, wu:fj43a08, zgc:111899, zgc:55483
Chromosome Location
Chr.24: 17364588 - 17400217 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213495.1 NP_998660.1 11 - 12 1561 123
XM_021470126.1 XP_021325801.1 11 - 12 1648 123
GenBank mRNA BC048370.1 - 11 - 12 1561 123
BC165368.1 - 10 - 11 1327 123

View Full Product Details