Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cul1b
Entrez Gene ID
406816
Gene Name
cullin 1b
Gene Aliases
cul1, wu:fi43h07, wu:fj43a08, zgc:111899, zgc:55483
Chromosome Location
Chr.24: 17364588 - 17400217 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213495.1 NP_998660.1 15 - 16 1939 80
XM_021470126.1 XP_021325801.1 15 - 16 2026 80
GenBank mRNA BC048370.1 - 15 - 16 1939 80
BC165368.1 - 14 - 15 1705 80

View Full Product Details