Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cul1b
Entrez Gene ID
406816
Gene Name
cullin 1b
Gene Aliases
cul1, wu:fi43h07, wu:fj43a08, zgc:111899, zgc:55483
Chromosome Location
Chr.24: 17364588 - 17400217 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213495.1 NP_998660.1 21 - 22 2506 86
GenBank mRNA BC048370.1 - 21 - 22 2506 86
BC165368.1 - 20 - 21 2272 86

View Full Product Details