Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cul1b
Entrez Gene ID
406816
Gene Name
cullin 1b
Gene Aliases
cul1, wu:fi43h07, wu:fj43a08, zgc:111899, zgc:55483
Chromosome Location
Chr.24: 17364588 - 17400217 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213495.1 NP_998660.1 4 - 5 743 144
XM_021470126.1 XP_021325801.1 4 - 5 830 144
GenBank mRNA BC048370.1 - 4 - 5 743 144
BC095589.1 - 4 - 5 756 144
BC165368.1 - 3 - 4 509 144

View Full Product Details