Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pcdh15a
Entrez Gene ID
368476
Gene Name
protocadherin-related 15a
Gene Aliases
PCDH15, si:bac7cse.1, si:xx-bac7cse.1
Chromosome Location
Chr.13: 41124593 - 41546792 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012482.1 NP_001012500.1 29 - 30 4278 99
XM_009307525.2 XP_009305800.1 29 - 30 4290 99
XM_009307527.2 XP_009305802.1 28 - 29 4275 99
XM_017358709.2 XP_017214198.1 29 - 30 4290 99
XM_017358710.2 XP_017214199.1 29 - 30 4290 99
XM_017358711.2 XP_017214200.1 29 - 30 4290 99
XM_017358712.2 XP_017214201.1 29 - 30 4290 99
XM_017358713.2 XP_017214202.1 29 - 30 4290 99
XM_017358714.2 XP_017214203.1 29 - 30 4290 99
GenBank mRNA AY772390.1 - 29 - 30 4278 99
KY432405.1 - 29 - 30 4277 99

View Full Product Details