Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pcdh15a
Entrez Gene ID
368476
Gene Name
protocadherin-related 15a
Gene Aliases
PCDH15, si:bac7cse.1, si:xx-bac7cse.1
Chromosome Location
Chr.13: 41124593 - 41546792 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012482.1 NP_001012500.1 17 - 18 2461 95
XM_009307525.2 XP_009305800.1 17 - 18 2473 95
XM_009307527.2 XP_009305802.1 16 - 17 2458 95
XM_017358709.2 XP_017214198.1 17 - 18 2473 95
XM_017358710.2 XP_017214199.1 17 - 18 2473 95
XM_017358711.2 XP_017214200.1 17 - 18 2473 95
XM_017358712.2 XP_017214201.1 17 - 18 2473 95
XM_017358713.2 XP_017214202.1 17 - 18 2473 95
XM_017358714.2 XP_017214203.1 17 - 18 2473 95
GenBank mRNA AY772390.1 - 17 - 18 2461 95
KY432405.1 - 17 - 18 2460 95

View Full Product Details