Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tlr3
Entrez Gene ID
403126
Gene Name
toll-like receptor 3
Gene Aliases
zgc:123006
Chromosome Location
Chr.1: 238623 - 243906 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013269.3 NP_001013287.2 4 - 5 2722 152
GenBank mRNA AY389446.1 - 1 - 2 315 152
BC107955.1 - 4 - 5 2726 152

View Full Product Details