Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ncs1b
Entrez Gene ID
553174
Gene Name
neuronal calcium sensor 1b
Gene Aliases
fa55b03, freqb, wu:fa55b03
Chromosome Location
Chr.8: 32512075 - 32599855 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020514.2 NP_001018350.1 4 - 5 867 130
XM_021478256.1 XP_021333931.1 5 - 6 1757 130
GenBank mRNA AY605665.1 - 4 - 5 322 130
BC116500.1 - 4 - 5 867 130

View Full Product Details