Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pip5k1aa
Entrez Gene ID
553629
Gene Name
phosphatidylinositol-4-phosphate 5-kinase, type I, alpha, a
Gene Aliases
pip5k1a, zgc:110576
Chromosome Location
Chr.19: 10892468 - 10915898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020602.1 NP_001018438.1 8 - 9 1164 81
GenBank mRNA BC095318.1 - 8 - 9 1164 81

View Full Product Details