Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il20ra
Entrez Gene ID
556728
Gene Name
interleukin 20 receptor, alpha
Gene Aliases
crfb8, im:7156745, si:ch211-272f15.1, zgc:158705
Chromosome Location
Chr.20: 2589400 - 2609276 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079676.2 NP_001073144.2 6 - 7 983 67
GenBank mRNA AM084416.2 - 6 - 7 933 67
EF014958.1 - 6 - 7 975 67

View Full Product Details