Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tnika
Entrez Gene ID
325042
Gene Name
TRAF2 and NCK interacting kinase a
Gene Aliases
fc52d11, msn, tnikl, wu:fc52d11, zgc:63535
Chromosome Location
Chr.2: 25658081 - 25815828 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199703.1 NP_955997.1 17 - 18 2339 70
XM_005163387.4 XP_005163444.1 19 - 20 2718 70
XM_005163388.4 XP_005163445.1 19 - 20 2700 70
XM_005163389.4 XP_005163446.1 18 - 19 2631 70
XM_005163390.4 XP_005163447.1 18 - 19 2457 70
XM_005163391.4 XP_005163448.1 18 - 19 2439 70
XM_005163392.4 XP_005163449.1 17 - 18 2352 70
XM_005163393.4 XP_005163450.1 4 - 5 843 70
GenBank mRNA BC055134.1 - 17 - 18 2339 70

View Full Product Details