Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- mgrn1a
- Entrez Gene ID
-
793939
- Gene Name
- mahogunin, ring finger 1a
- Gene Aliases
-
mgrn1, wu:fi38c03, zgc:55978
- Chromosome Location
-
Chr.12: 20122461 - 20147764 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC048069.1 |
- |
- |
1770 |
67 |
View Full Product Details