Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lman2la
Entrez Gene ID
403038
Gene Name
lectin, mannose-binding 2-like a
Gene Aliases
VIP36-like, lman2l, zgc:113949
Chromosome Location
Chr.5: 72377823 - 72383840 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205725.2 NP_991288.2 2 - 3 262 98
GenBank mRNA AF506214.1 - 2 - 3 297 98
BC097031.1 - 2 - 3 262 98

View Full Product Details