Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pak5
Entrez Gene ID
405898
Gene Name
p21 protein (Cdc42/Rac)-activated kinase 5
Gene Aliases
pak6, pak7, zgc:76890
Chromosome Location
Chr.20: 54689312 - 54744600 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212962.1 NP_998127.1 2 - 3 1587 112
GenBank mRNA BC068016.1 - 2 - 3 1587 112
BC168506.1 - 2 - 3 1482 112

View Full Product Details