Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
camk2n2
Entrez Gene ID
436915
Gene Name
calcium/calmodulin-dependent protein kinase II inhibitor 2
Gene Aliases
zgc:92198
Chromosome Location
Chr.24: 25692802 - 25695057 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002642.1 NP_001002642.1 1 - 2 389 99
GenBank mRNA BC075931.1 - 1 - 2 389 99

View Full Product Details