Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nosip
Entrez Gene ID
492793
Gene Name
nitric oxide synthase interacting protein
Gene Aliases
wu:fb80a11, zgc:101669
Chromosome Location
Chr.3: 19662 - 23395 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007434.1 NP_001007435.1 1 - 2 341 75
XM_005164022.4 XP_005164079.1 2 - 3 181 75
XM_009299536.3 XP_009297811.1 2 - 3 239 75
GenBank mRNA BC085409.1 - 1 - 2 341 75
BC164074.1 - 1 - 2 100 75

View Full Product Details