The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045232.1 | NP_001038697.1 | 1 - 1 | 746 | 86 |
XM_005164771.4 | XP_005164828.1 | 2 - 2 | 848 | 86 |