Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tfam
Entrez Gene ID
571106
Gene Name
transcription factor A, mitochondrial
Gene Aliases
zgc:153358
Chromosome Location
Chr.12: 7254112 - 7263958 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077389.1 NP_001070857.1 6 - 7 753 89
GenBank mRNA BC124667.1 - 6 - 7 753 89
BC165544.1 - 6 - 7 625 89

View Full Product Details