Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stk17b
Entrez Gene ID
393507
Gene Name
serine/threonine kinase 17b (apoptosis-inducing)
Gene Aliases
zgc:65989
Chromosome Location
Chr.9: 306390 - 314498 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200535.1 NP_956829.1 3 - 4 489 88
GenBank mRNA BC056298.1 - 3 - 4 489 88

View Full Product Details