Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klf6a
Entrez Gene ID
280650
Gene Name
Kruppel-like factor 6a
Gene Aliases
cb538, copeb, klf6, wu:fb76d04, wu:fb92h03, wu:fe26g06, wu:fk78f12
Chromosome Location
Chr.24: 4141479 - 4148914 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201461.2 NP_958869.2 3 - 4 1032 120
GenBank mRNA BC048893.1 - 3 - 4 990 120
CT734412.2 - 2 - 3 360 120

View Full Product Details