Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab5b
Entrez Gene ID
405821
Gene Name
RAB5B, member RAS oncogene family
Gene Aliases
zgc:76978
Chromosome Location
Chr.9: 417480 - 430312 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212885.1 NP_998050.1 3 - 4 467 71
GenBank mRNA BC066634.1 - 3 - 4 467 71

View Full Product Details