Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stx5a
Entrez Gene ID
436605
Gene Name
syntaxin 5A
Gene Aliases
wu:fj81f08, zgc:91864
Chromosome Location
Chr.14: 216499 - 236885 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002333.1 NP_001002333.1 5 - 6 517 74
GenBank mRNA BC076467.1 - 5 - 6 517 74

View Full Product Details