Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab24
Entrez Gene ID
574423
Gene Name
RAB24, member RAS oncogene family
Gene Aliases
im:7142760, im:7155843, zgc:112047
Chromosome Location
Chr.21: 38732976 - 38740009 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029970.2 NP_001025141.1 6 - 7 909 70
GenBank mRNA BC098610.1 - 6 - 7 842 70
EH548684.1 - 6 - 7 906 70
EH563661.1 - 3 - 4 350 70

View Full Product Details