Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scrn2
Entrez Gene ID
558306
Gene Name
secernin 2
Gene Aliases
si:ch211-184m19.2
Chromosome Location
Chr.20: 14635428 - 14680897 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030116.1 NP_001025287.1 6 - 7 962 78
XM_005160433.1 XP_005160490.1 6 - 7 1018 78
XM_005160434.1 XP_005160491.1 6 - 7 1018 78
GenBank mRNA BC124746.1 - 6 - 7 965 78

View Full Product Details