Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
caspb
Entrez Gene ID
259303
Gene Name
caspase b
Gene Aliases
casp19a, caspy2, si:ch211-233g6.8, zgc:109807
Chromosome Location
Chr.1: 58050910 - 58064721 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152884.2 NP_690840.2 3 - 4 521 100
GenBank mRNA AF327410.1 - 2 - 3 484 100
BC095000.1 - 3 - 4 521 100
BC164498.1 - 2 - 3 518 100
DQ022755.1 - 2 - 3 100
MG958005.1 - 2 - 3 484 100

View Full Product Details